missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, anhydrous, 97%

Product Code. 11398708
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398708 1 kg 1kg
11378708 250 g 250g
11388708 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398708 Supplier Thermo Scientific Alfa Aesar Supplier No. 041587.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-53-2 | K3O4P | 212.265 g/mol

Potassium phosphate is used as a fertilizer and food additive. It serves as a buffering agent to regulate pH. It is also used in optical modulators and for non-linear optics such as second-harmonic generation (SHG). It is involved in the phase-transfer catalyzed preparation of acetylenes from the corresponding vicinal dibromo compounds through double dehydrobromination. It is also useful for the oxidative coupling of thiols to disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.265
MDL Number MFCD00036295
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
Synonym potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
PubChem CID 62657
IUPAC Name tripotassium;phosphate
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340°C
Physical Form Granules
Quantity 1 kg
Assay Percent Range 97%
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 212.27
Sensitivity Hygroscopic
Density 2.564 g/mL
Chemical Name or Material Potassium phosphate, anhydrous
compliance-icons compliance-icons
Product Identifier
  • Potassium phosphate, anhydrous
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Potassium phosphate,anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.