missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, 97%

Product Code. 15489019
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2000g
500g
Product Code. Quantity unitSize
15489019 500 g 500g
15479019 100 g 100g
15469019 2000 g 2000g
3 options
This item is not returnable. View return policy

Product Code. 15489019

Brand: Thermo Scientific Alfa Aesar L15168.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-53-2 | K{3}O{4}P

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 1340°C
CAS 7778-53-2
Physical Form Solid
Quantity 500 g
Molecular Formula K{3}O{4}P
Synonym Potassium phosphate, tribasic;Tripotassium phosphate
Percent Purity 97%
compliance-icons compliance-icons
Product Identifier
  • Potassium phosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Phosphoric acid, tripotassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.