missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, 0.5M buffer soln., pH 7.6, Thermo Scientific™
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline. In addition to helping maintain pH, it supplies essential phosphate.
Solubility
Soluble in water, Insoluble in ethanol, alcohol.
Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline. In addition to helping maintain pH, it supplies essential phosphate.
Solubility
Soluble in water, Insoluble in ethanol, alcohol.
Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
Specifications
Specifications
| CAS | 7778-77-0 |
| Concentration | 0.5 M |
| pH | 7.6 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Molecular Formula | H2KO4P |
| MDL Number | MFCD00011401 MFCD00147253 |
| InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
| SMILES | [K+].OP(O)([O-])=O |
| IUPAC Name | potassium dihydrogen phosphate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction