Learn More
Thermo Scientific Chemicals Potassium phosphate, 0.5M buffer soln., pH 7.0
Description
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.
Solubility
Soluble in water, Insoluble to slightly soluble in ethanol, insoluble in alcohol.
Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
Specifications
Specifications
| Concentration | 0.5 M |
| pH | 7.0 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Chemical Name or Material | Potassium phosphate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.