missing translation for 'onlineSavingsMsg'
Learn More

Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Product Code. 15494697
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494697 1 g 1g
15404707 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494697 Supplier Thermo Scientific Alfa Aesar Supplier No. H32030.03
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1027642-30-3 | Molecular Formula: C7H7BF3KO | Molecular Weight: 214.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1027642-30-3
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD10566516
InChI Key OVYSNXCRBZPJIG-UHFFFAOYSA-N
Synonym potassium phenoxy-methyltrifluoroborate, potassium trifluoro phenoxymethyl boranuide, pubchem11600, potassium phenoxymethyltrifluoroborate, potassium trifluoro phenoxymethyl borate, potassium trifluoro phenoxymethyl borate 1-, potassium phenoxymethyltrifluoroborate, contains kbr
PubChem CID 45479874
IUPAC Name potassium;trifluoro(phenoxymethyl)boranuide
SMILES [B-](COC1=CC=CC=C1)(F)(F)F.[K+]

Specifications

Quantity 1 g
Formula Weight 214.04
Percent Purity 95%
Chemical Name or Material Potassium phenoxymethyltrifluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.