missing translation for 'onlineSavingsMsg'
Learn More

Potassium Persulfate, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Product Code. 15578094 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15578094 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578094 Supplier Avantor J.T.Baker Supplier No. 323901

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    Suitable for Phosphate and Mercury Determinations

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.309
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
PubChem CID 24412
IUPAC Name dipotassium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]

Specifications

Quantity 500 g
Formula Weight 270.32
Percent Purity 99.5%
Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.48kg
Chemical Name or Material Potassium Persulfate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.