missing translation for 'onlineSavingsMsg'
Learn More

Potassium persulfate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10646062
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.3
MDL Number MFCD00011386
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
PubChem CID 24412
IUPAC Name dipotassium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 100.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99% min. (Iodometry)
Linear Formula K2S2O8
Merck Index 15,7777
Solubility Information Solubility in water: 5g/100mL (20°C). Other solubilities: insoluble in alcohol
Formula Weight 270.3
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 2.47
Chemical Name or Material Potassium persulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'