missing translation for 'onlineSavingsMsg'
Learn More

Potassium perruthenate(VII), 97%

Product Code. 11351958
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351958 5 g 5g
11361958 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351958 Supplier Thermo Scientific Alfa Aesar Supplier No. 011877.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10378-50-4 | C32H36N2O5 | 528.65 g/mol

Potassium perruthenate(VII) is used in the production of carbon nanotubes for flexible solid-state supercapacitors. It is also used as an oxidizing agent in the synthesis of 5-formylcytosine from 5-hydroxymethylcytosine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-50-4
Molecular Formula C32H36N2O5
Molecular Weight (g/mol) 528.65
MDL Number MFCD00061333
InChI Key PXMOBROHNZPLTM-UHFFFAOYSA-N
Synonym 4,7-bis benzyloxy-n,n-diisopropyl-1,3-dioxo-2-isoindolinebutyramide
PubChem CID 101141422
IUPAC Name 4-[1,3-dioxo-4,7-bis(phenylmethoxy)isoindol-2-yl]-N,N-di(propan-2-yl)butanamide
SMILES CC(C)N(C(C)C)C(=O)CCCN1C(=O)C2=C(OCC3=CC=CC=C3)C=CC(OCC3=CC=CC=C3)=C2C1=O

Specifications

Melting Point 440°C (decomposition)
Color Black
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 97%
Solubility Information Slightly soluble in water.
Formula Weight 204.17
Appearance Black crystals
Chemical Name or Material Potassium perruthenate(VII)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.