missing translation for 'onlineSavingsMsg'
Learn More

Potassium perrhenate, 99% (metals basis), Re 64%

Product Code. 11381219
Change view
Click to view available options
Quantity:
0.2 g
2 g
10 g
50 g
Unit Size:
0.2g
10g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381219 10 g 10g
11391219 0.2 g 0.2g
11371219 2 g 2g
11301229 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11381219 Supplier Thermo Scientific Alfa Aesar Supplier No. 011411.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A powerful oxidizer | CAS: 10466-65-6 | KO4Re | 289.30 g/mol

A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 10466-65-6
Molecular Formula KO4Re
Molecular Weight (g/mol) 289.30
MDL Number MFCD00011365
InChI Key QFKRWIFGDGKWLM-UHFFFAOYSA-N
Synonym potassium perrhenate, potassiumperrhenate, k.o4re, potassium rhenium oxide kreo4, perrhenic acid hreo4 , potassium salt, potassium perrhenate vii, acmc-1bx96, perrhenic acid potassium salt, potassium oxido trioxo rhenium, potassium perrhenate kreo4
PubChem CID 3661560
IUPAC Name potassium;oxido(trioxo)rhenium
SMILES [K+].[O-][Re](=O)(=O)=O

Tekniske data

Boiling Point 1360°C to 1370°C
Melting Point 550°C
Color White
Physical Form Crystalline
Quantity 10 g
UN Number UN1479
Solubility Information Soluble in water.
Formula Weight 289.3
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Re 64%
Odor Odorless
Refractive Index 1.643
Density 4.887 g/mL
Assay (metals basis)
Chemical Name or Material Potassium perrhenate
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.