missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium peroxydisulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
Potassium persulfate is a powerful oxidant, commonly used to initiate polymerizations. Potassium Persulphate used for bleaching and textile desizing, as an oxidizing agent and antiseptic, in purification of ammonium sulfate, and in the manufacture of soap and pharmaceuticals. It is also used as al laboratory oxidant and photography chemical. It is a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 100°C (dec.) |
Color | White |
Physical Form | Crystalline |
Quantity | 1 kg |
Linear Formula | K2S2O8 |
UN Number | UN1492 |
Merck Index | 14,7656 |
Solubility Information | It is highly soluble in water. It is insoluble in alcohol. |
Formula Weight | 270.31 |
Percent Purity | ≥99.0% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'