missing translation for 'onlineSavingsMsg'
Learn More

Potassium peroxydisulfate, Electrophoresis Grade, 99.0%

Product Code. 15440627
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440627 100 g 100g
15420627 5 g 5g
15430627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440627 Supplier Thermo Scientific Alfa Aesar Supplier No. J66672.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7727-21-1 | K2O8S2 | 270.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.309
MDL Number MFCD00011386
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
PubChem CID 24412
IUPAC Name dipotassium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 100°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
UN Number UN1492
Merck Index 14,7656
Formula Weight 270.31
Percent Purity 99.0%
Grade Electrophoresis
Odor Odorless
Density 2.477 g/mL
Chemical Name or Material Potassium peroxydisulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.