missing translation for 'onlineSavingsMsg'
Learn More

Potassium peroxydisulfate, 97%

Product Code. 11488186
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488186 2500 g 2500g
11478186 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488186 Supplier Thermo Scientific Alfa Aesar Supplier No. A15310.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7727-21-1 | K2O8S2 | 270.309 g/mol

Potassium peroxydisulfate is used as an oxidizing agent in organic synthesis. It is involved in the Elbs persulfate oxidation of phenols and the Boyland-Sims oxidation of anilines. In solution, it gives radicals and is used to initiate polymerization reactions to prepare styrene-butadiene rubber and polytetrafluoroethylene. It is used as a bleaching agent in various hair bleaches, as an ion exchange agent, a plating agent and a surface treating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium peroxydisulfate is used as an oxidizing agent in organic synthesis. It is involved in the Elbs persulfate oxidation of phenols and the Boyland-Sims oxidation of anilines. In solution, it gives radicals and is used to initiate polymerization reactions to prepare styrene-butadiene rubber and polytetrafluoroethylene. It is used as a bleaching agent in various hair bleaches, as an ion exchange agent, a plating agent and a surface treating agent.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Moisture sensitive. Incompatible with organic materials, strong reducing agents, powdered metals, strong bases, alcohols, phosphorous, anhydrides, halogens and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.309
MDL Number MFCD00011386
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
PubChem CID 24412
IUPAC Name dipotassium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 100°C (dec.)
Quantity 2500 g
Odor Odorless
Density 2.477
Linear Formula K2S2O8
UN Number UN1492
Merck Index 14,7656
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 270.33
Percent Purity 97%
Chemical Name or Material Potassium peroxydisulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium peroxydisulfate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.