missing translation for 'onlineSavingsMsg'
Learn More

Potassium permanganate, Standard solution for volumetric analysis, solution 0.02M (0.1N), meets spec. of Ph. Eur., Thermo Scientific™

Product Code. 17790719 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Appearance:
Purple-black liquid
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Appearance Unit Size
17790719 Purple-black liquid 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 17790719 Supplier Thermo Scientific Acros Supplier No. 474370010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Specifications

Chemical Name or Material Potassium permanganate
CAS 7722-64-7
Linear Formula KMnO4
Merck Index XIII 7740
Product Line Reagents
Appearance Purple-black liquid
Density 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.