missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate solution 0.01mol/L, Fisher Chemical™
BIOPHARMA

Product Code. 12933594 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12933594 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 12933594 Supplier Fisher Chemical Supplier No. VL 276 1000
  • ONLINE EXCLUSIVE PRICE
    £78.15
    £72.88 / 1000mL
    Save £5.27
    6% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7722-64-7, 7732-18-5
Molecular Formula KMnO4
Concentration 0.01 M
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES [K+].[O-][Mn](=O)(=O)=O
IUPAC Name potassium manganesoylolate
Molecular Weight (g/mol) 158.03
PubChem CID 516875
Quantity 1000 mL
Chemical Name or Material Potassium Permanganate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.