missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate, puriss p.a, ACS, ≥99.0% (RT), Honeywell Fluka™
Puriss. p.a., ACS Reagent, Hg ≤0.000005%,≥99.0% (RT)
Chemical Identifiers
CAS | 7722-64-7 |
---|---|
Molecular Formula | KMnO4 |
Molecular Weight (g/mol) | 158.032 |
MDL Number | MFCD00011364 |
InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
PubChem CID | 516875 |
IUPAC Name | potassium;permanganate |
SMILES | [O-][Mn](=O)(=O)=O.[K+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7722-64-7 | |
158.032 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
516875 | |
[O-][Mn](=O)(=O)=O.[K+] |
KMnO4 | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
potassium;permanganate |
Specifications
7722-64-7 | |
Crystalline | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
[O-][Mn](=O)(=O)=O.[K+] | |
158.032 | |
158.03g/mol | |
Odorless | |
2.7g/cm3 |
Violet | |
KMnO4 | |
UN1490 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
potassium;permanganate | |
516875 | |
≥99.0% (RT) | |
>240°C | |
Potassium permanganate |
Safety and Handling
P220-P273-P501
missing translation for 'dotInformation' : 5.1 / PGII
missing translation for 'rtecsNumber' : 231-760-3