Learn More
Potassium permanganate, low in mercury, ACS, 99.0% min, Thermo Scientific™
£24.50 - £217.00
Chemical Identifiers
| CAS | 7722-64-7 |
|---|---|
| Molecular Formula | KMnO4 |
| Molecular Weight (g/mol) | 158.032 |
| MDL Number | MFCD00011364 |
| InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
| Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
| PubChem CID | 516875 |
| IUPAC Name | potassium;permanganate |
| SMILES | [O-][Mn](=O)(=O)=O.[K+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11324197
|
Thermo Scientific Alfa Aesar
036675.22 |
100g |
£24.50
100g |
Estimated Shipment: 22-07-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11304197
|
Thermo Scientific Alfa Aesar
036675.36 |
500g |
£65.40
500g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11314197
|
Thermo Scientific Alfa Aesar
036675.A4 |
2.5kg |
£217.00
2.5kg |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| KMnO4 | |
| MFCD00011364 | |
| potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
| potassium;permanganate |
Specifications
| 7722-64-7 | |
| KMnO4 | |
| potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
| VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
| potassium;permanganate | |
| 516875 | |
| ACS | |
| Crystalline | |
| Potassium permanganate, For analysis ACS |
| 99% min. | |
| MFCD00011364 | |
| Soluble in water. | |
| [O-][Mn](=O)(=O)=O.[K+] | |
| 158.032 | |
| 158.03 | |
| Light sensitive | |
| 2.7 |
Safety and Handling
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H272-H302-H314-H335-H361d-H373
missing translation for 'dotInformation' : DOT Class: 5.1; Packing Group: II
missing translation for 'einecsNumber' : 231-760-3
missing translation for 'rtecsNumber' : SD6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.