missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate, low in mercury, ACS, 99.0% min, Thermo Scientific™
Click to view available options
Quantity:
100g
500g
2.5kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 2.5kg |
Assay Percent Range | 99% min. |
Solubility Information | Soluble in water. |
Formula Weight | 158.03 |
Grade | ACS |
Sensitivity | Light sensitive |
Appearance | Crystalline |
Density | 2.7 |
Chemical Name or Material | Potassium permanganate, For analysis ACS |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'