missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate Concentrate, 1/50M, For 1L standard solution, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Color | Violet |
| Physical Form | Liquid |
| Quantity | 6 x 1 Ea. |
| Packaging | Ampoule |
| Appearance | Clear liquid |
| UN Number | UN3214 |
| Formula Weight | 158.03g/mol |
| Concentration or Composition (by Analyte or Components) | 1/50 M KMnO4 (0.1N) |
| Chemical Name or Material | Potassium permanganate concentrate, For 1L standard solution, 1/50M KMnO4 (0.1N) |
Safety and Handling
Product Identifier
- Potassium permanganate concentrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: potassium permanganate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?