missing translation for 'onlineSavingsMsg'
Learn More

Potassium permanganate, 98%, Thermo Scientific™

£41.30 - £341.00


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11484803
View Documents
Thermo Scientific Alfa Aesar
A12170.0C
£341.00
10000g
View alternative products
Save Up to 
»
Estimated Shipment: 22-07-2026
Add to basket
 
 Restricted Item
11464803
View Documents
Thermo Scientific Alfa Aesar
A12170.36
500g
£41.30
500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11474803
View Documents
Thermo Scientific Alfa Aesar
A12170.0E
2500g
£117.00
2500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.

Solubility
Soluble in water.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, peroxides, zinc, copper, alcohols, hydrogen fluoride and acids.
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.