Learn More
Potassium permanganate, 0.1N Standardized Solution, Thermo Scientific™
Specifications
| CAS | 7722-64-7 |
|---|---|
| Molecular Formula | KMnO4 |
| Molecular Weight (g/mol) | 158.032 |
| MDL Number | MFCD00011364 |
| InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H361d
missing translation for 'einecsNumber' : 231-760-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.