missing translation for 'onlineSavingsMsg'
Learn More

Potassium perchlorate, 99%

Product Code. 11453674
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11453674

Brand: Thermo Scientific Alfa Aesar A11296.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7778-74-7 | ClKO4 | 138.54 g/mol

Potassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.

Solubility
Soluble in water. Slightly soluble in acetone. Insoluble in ethanol, ethyl acetate and ether.

Notes
Hygroscopic. Incompatible with strong reducing agents, powdered metals, strong acids and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-74-7
Molecular Formula ClKO4
Molecular Weight (g/mol) 138.54
MDL Number MFCD00011362
InChI Key YLMGFJXSLBMXHK-UHFFFAOYSA-M
Synonym potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
PubChem CID 516900
SMILES [K+].[O-][Cl](=O)(=O)=O

Specifications

Melting Point 301°C to 304°C
Quantity 100 g
UN Number UN1489
Merck Index 14,7653
Solubility Information Soluble in water. Slightly soluble in acetone. Insoluble in ethanol,ethyl acetate and ether.
IUPAC Name potassium;perchlorate
Formula Weight 138.55
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.52
Chemical Name or Material Potassium perchlorate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium perchlorate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium perchlorate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.