missing translation for 'onlineSavingsMsg'
Learn More

Potassium perchlorate, 99+%, for analysis

Product Code. 10410881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10410881 500 g Plastic bottle 500g
10286320 25 g Plastic bottle 25g
10075363 1 kg Plastic bottle 1kg
13438478 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10410881 Supplier Thermo Scientific Acros Supplier No. 221365000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7778-74-7 | ClKO4 | 138.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-74-7
Molecular Formula ClKO4
Molecular Weight (g/mol) 138.54
MDL Number MFCD00011362
InChI Key YLMGFJXSLBMXHK-UHFFFAOYSA-M
Synonym potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
PubChem CID 516900
IUPAC Name potassium;perchlorate
SMILES [K+].[O-][Cl](=O)(=O)=O

Specifications

Melting Point 400.0°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 500 g
Assay Percent Range 99+%
Linear Formula KClO4
Merck Index 15,7774
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: 0.75 g/100mL water (0°C), 21.8 g/100mL water (100°C), practically insoluble in alcohol
Formula Weight 138.55
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium perchlorate, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.