missing translation for 'onlineSavingsMsg'
Learn More

Potassium pentachlororuthenate(III) hydrate, 99.95% (metals basis), Ru 25.4% min

Product Code. 11326558
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326558 5 g 5g
11336558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326558 Supplier Thermo Scientific Alfa Aesar Supplier No. 012173.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14404-33-2 | Cl5K2Ru | 356.52 g/mol

Potassium pentachlororuthenate(III) hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14404-33-2
Molecular Formula Cl5K2Ru
Molecular Weight (g/mol) 356.52
MDL Number MFCD00058747
InChI Key IXYMQZXAJLWXRK-UHFFFAOYSA-I
Synonym dipotassium aquapentachlororuthenate, potassium pentachlororuthenate iii hydrate, dipotassium aquapentachlororuthenate iii, dipotassium pentachlororutheniumdiuide hydrate, potassium aquapentachlororuthenate iii , premion, ruthenate 2-, aquapentachloro-, dipotassium, oc-6-21
PubChem CID 16211344
IUPAC Name dipotassium;pentachlororuthenium(2-);hydrate
SMILES [K+].[K+].Cl[Ru--](Cl)(Cl)(Cl)Cl

Specifications

Physical Form Crystalline Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 356.54 (Anhydrous)
Percent Purity 99.95%
Concentration or Composition (by Analyte or Components) Ru 25.4% min.
Assay (metals basis)
Chemical Name or Material Potassium pentachlororuthenate(III) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.