missing translation for 'onlineSavingsMsg'
Learn More

Potassium pentachlororhodate(III), Premion™, 99.99% (metals basis), Rh 28.2% min, Thermo Scientific Chemicals

Product Code. 11364008
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364008 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11364008 Supplier Thermo Scientific Alfa Aesar Supplier No. 040409.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65980-75-8
Molecular Formula Cl5K2Rh
Molecular Weight (g/mol) 358.352
MDL Number MFCD00049661
InChI Key YEPORAXCNSGGEV-UHFFFAOYSA-I
Synonym potassium pentachlororhodate iii, pentachlororhodate 2-dipotassium, ccris 7830, potassium pentachlororhodate 2-, dipotassium pentachlororhodate 2-, dipotassium pentachlororhodate, rhodate 2-, pentachloro-, dipotassium, dipotassium pentachlororhodiumdiuide, cl5rh.2k, rhodate 2-, pentachloro-, potassium 1:2
PubChem CID 149285
IUPAC Name dipotassium;pentachlororhodium(2-)
SMILES Cl[Rh-2](Cl)(Cl)(Cl)Cl.[K+].[K+]

Specifications

Physical Form Powder
Quantity 1 g
Solubility Information Slightly soluble in cold water; Insoluble in alcohol
Formula Weight 358.37
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Rh 28.2% min.
Assay (metals basis)
Chemical Name or Material Potassium pentachlororhodate(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.