Learn More
Potassium pentachlororhodate(III), Premion™, 99.99% (metals basis), Rh 28.2% min, Thermo Scientific Chemicals
£648.00
Chemical Identifiers
CAS | 65980-75-8 |
---|---|
Molecular Formula | Cl5K2Rh |
Molecular Weight (g/mol) | 358.352 |
MDL Number | MFCD00049661 |
InChI Key | YEPORAXCNSGGEV-UHFFFAOYSA-I |
Synonym | potassium pentachlororhodate iii, pentachlororhodate 2-dipotassium, ccris 7830, potassium pentachlororhodate 2-, dipotassium pentachlororhodate 2-, dipotassium pentachlororhodate, rhodate 2-, pentachloro-, dipotassium, dipotassium pentachlororhodiumdiuide, cl5rh.2k, rhodate 2-, pentachloro-, potassium 1:2 |
PubChem CID | 149285 |
IUPAC Name | dipotassium;pentachlororhodium(2-) |
SMILES | Cl[Rh-2](Cl)(Cl)(Cl)Cl.[K+].[K+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65980-75-8 | |
358.352 | |
YEPORAXCNSGGEV-UHFFFAOYSA-I | |
149285 | |
Cl[Rh-2](Cl)(Cl)(Cl)Cl.[K+].[K+] |
Cl5K2Rh | |
MFCD00049661 | |
potassium pentachlororhodate iii, pentachlororhodate 2-dipotassium, ccris 7830, potassium pentachlororhodate 2-, dipotassium pentachlororhodate 2-, dipotassium pentachlororhodate, rhodate 2-, pentachloro-, dipotassium, dipotassium pentachlororhodiumdiuide, cl5rh.2k, rhodate 2-, pentachloro-, potassium 1:2 | |
dipotassium;pentachlororhodium(2-) |
Specifications
65980-75-8 | |
Cl5K2Rh | |
potassium pentachlororhodate iii, pentachlororhodate 2-dipotassium, ccris 7830, potassium pentachlororhodate 2-, dipotassium pentachlororhodate 2-, dipotassium pentachlororhodate, rhodate 2-, pentachloro-, dipotassium, dipotassium pentachlororhodiumdiuide, cl5rh.2k, rhodate 2-, pentachloro-, potassium 1:2 | |
YEPORAXCNSGGEV-UHFFFAOYSA-I | |
dipotassium;pentachlororhodium(2-) | |
149285 | |
99.99% | |
Rh 28.2% min. | |
Potassium pentachlororhodate(III) |
Powder | |
MFCD00049661 | |
Slightly soluble in cold water; Insoluble in alcohol | |
Cl[Rh-2](Cl)(Cl)(Cl)Cl.[K+].[K+] | |
358.352 | |
358.37 | |
Premion™ | |
(metals basis) |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
missing translation for 'einecsNumber' : 265-989-5
missing translation for 'rtecsNumber' : VI8580332
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only