missing translation for 'onlineSavingsMsg'
Learn More

Potassium pentachloronitrosylruthenate(II), Ru 25.8%, Thermo Scientific Chemicals

Product Code. 11336048
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as precipitation agent in Heterobimetallic coordination polymers involving 3d metal complexes and heavier transition metals cyanometallates. Ruthenium complexes are presently an object of great attention in the field of medicinal chemistry, as antitumor agents with selective antimetastatic properties and low systemic toxicity. Ruthenium compounds appear to penetrate reasonably well the tumor cells and bind effectively to DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14854-54-7
Molecular Formula Cl5K2NORu
Molecular Weight (g/mol) 386.52
MDL Number MFCD00049789
InChI Key ZLWZLQOGXCFPCK-UHFFFAOYSA-I
Synonym dipotassium nitrosylpentachlororuthenate ii, dipotassium pentachloro nitrosyl ruthenate ii, dipotassium pentachloro nitroso rutheniumdiuide, potassium pentachloronitrosylruthenate ii , ru
PubChem CID 71311320
SMILES [K+].[K+].Cl[Ru++](Cl)(Cl)(Cl)(Cl)N=O

Specifications

Melting Point 220°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name dipotassium pentachloro(nitroso)rutheniumbis(ylium)
Formula Weight 386.55
Concentration or Composition (by Analyte or Components) Ru 25.8%
Chemical Name or Material Potassium pentachloronitrosylruthenate(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'