missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium p-Nitrophenyl Sulphate, TRC
Specifications
Specifications
| CAS | 6217-68-1 |
| Molecular Formula | C6 H4 N O6 S . K |
| Quantity | 50 mg |
| Synonym | 4-Nitrophenyl sulfate potassium, Sulfuric acid, mono(4-nitrophenyl) ester, potassium salt (1:1) (ACI), Phenol, p-nitro-, hydrogen sulfate (ester), potassium salt (8CI), Phenol, p-nitro-, hydrogen sulfate, potassium salt (6CI, 7CI), Sulfuric acid, mono(4-nitrophenyl) ester, potassium salt (9CI), Potassium 4-nitrophenyl sulfate, Potassium p-nitrophenyl sulfate |
| InChI Formula | InChI=1S/C6H5NO6S.K/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;/h1-4H,(H,10,11,12);/q;+1/p-1 |
| SMILES | [K+].[O-][N+](=O)c1ccc(OS(=O)(=O)[O-])cc1 |
| IUPAC Name | potassium;(4-nitrophenyl) sulfate |
| Molecular Weight (g/mol) | 257.26 |
| Formula Weight | 256.9396 |
| Chemical Name or Material | 4-Nitrophenyl sulfate potassium salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?