missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate Monohydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10417270 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10417270 500 g 500g
10385760 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10417270 Supplier Fisher Chemical Supplier No. P/6320/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 356°C
Color White
pH 7.0 to 8.5
Physical Form Solid
Quantity 500 g
IUPAC Name dipotassium hydrate oxalate
Formula Weight 184.24g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Potassium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.