missing translation for 'onlineSavingsMsg'
Learn More

Potassium osmium(VI) oxide dihydrate, 98+%

Product Code. 11377487
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377487 0.25 g 0.25g
11367487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377487 Supplier Thermo Scientific Alfa Aesar Supplier No. 012647.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10022-66-9 | H4K2O6Os | 368.453 g/mol

Potassium osmium(VI) oxide dihydrate acts as a catalyst for the oxidative cleavage of olefins to ketones and carboxylic acids. It is employed in an intramolecular amidohydroxylation of carbamoyloxytethered olefins providing hydroxy oxazolidinones. It is also used as a reagent in the synthesis of amphidinolide B, which shows potent antitumor activity against human solid and blood tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-66-9
Molecular Formula H4K2O6Os
Molecular Weight (g/mol) 368.453
MDL Number MFCD00149919
InChI Key DGODWNOPHMXOTR-UHFFFAOYSA-N
Synonym potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate
PubChem CID 53393272
IUPAC Name dipotassium;dioxido(dioxo)osmium;dihydrate
SMILES O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]

Specifications

Color White
Physical Form Powder
Quantity 0.25 g
Assay Percent Range 99+%
Solubility Information Soluble in water. Slowly decomposes in aqueous solution forming the tetroxide. Insoluble in alcohol,etherSoluble in water. Insoluble in alcohol and ether.
Formula Weight 368.43 (332.40 Anhydrous)
Sensitivity Hygroscopic
Appearance Purple powder
Chemical Name or Material Potassium osmium(VI) oxide dihydrate
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P405-Store locked up.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.