missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrite, meets analytical spec. of FCC, 97-100.5%, Honeywell Fluka™

Product Code. 15651180 Shop All Honeywell Fluka Products
This item is not returnable. View return policy
Product Code. 15651180 Supplier Honeywell-Fluka Supplier No. 1265450KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets analytical specification of FCC, 97-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 350°C (decomposition)(literature)
CAS 7758-09-0
Quantity 50 kg
Assay Percent Range 97 to 100.5%
Molecular Formula KNO2
Linear Formula KNO2
MDL Number MFCD00011408
UN Number UN1488
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
Solubility Information 0.35 parts Soluble in water, alcohol: slightly soluble
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES [K+].[O-]N=O
IUPAC Name potassium;nitrite
Molecular Weight (g/mol) 85.10
PubChem CID 516910
Formula Weight 85.1
Percent Purity 97 to 100.5%
Grade FCC
Packaging Fiberboard Box
Density 1.920 g/cm3
Chemical Name or Material Potassium nitrite, Meets analytical specification of FCC, 97-100.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.