missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, Extra Pure, SLR, Fisher Chemical

Product Code. 10627422 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10627422 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627422 Supplier Fisher Chemical Supplier No. P/6160/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 387°C
CAS 7758-09-0
Color White
pH 7 to 10
Physical Form Solid
Quantity 500 g
Molecular Formula KNO2
MDL Number MFCD00011408
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES [K+].[O-]N=O
IUPAC Name potassium;nitrite
Molecular Weight (g/mol) 85.10
PubChem CID 516910
Formula Weight 85.1g/mol
Packaging Plastic powder jar
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Potassium nitrite
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
Indicaciones de peligro
  • H400-Very toxic to aquatic life.
  • H301-Toxic if swallowed.
  • H272-May intensify fire; oxidiser.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.