missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, Certified AR for Analysis, Fisher Chemical

Product Code. 10386430 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10386430 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10386430 Supplier Fisher Chemical Supplier No. P/6162/50
Quantity Price
16 £101.67 / 250g
1 £114.22 / 250g
4 £107.01 / 250g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

250g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 387°C
CAS 7758-09-0
Color White
pH 7 to 10
Physical Form Solid
Quantity 250 g
Molecular Formula KNO2
MDL Number MFCD00011408
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES [K+].[O-]N=O
IUPAC Name potassium;nitrite
Molecular Weight (g/mol) 85.10
PubChem CID 516910
Formula Weight 85.1g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium nitrite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H301-Toxic if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.