missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrite, ACS, 96.0% min

Product Code. 11367817
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367817 10 g 10g
11347817 100 g 100g
11357817 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367817 Supplier Thermo Scientific Alfa Aesar Supplier No. 014245.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-09-0 | KNO2 | 85.10 g/mol

In analytical chemistry (testing for amino acids, cobalt, iodine, urea).Potassium nitrite is used as a food preservative, paint and coating additives and analytical reagent. It is used as a strong oxidizing agent as well as involved in the preparation of heat transfer salts. In addition, it is employed as corrosion inhibitors and anti-scaling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
IUPAC Name potassium;nitrite
SMILES [K+].[O-]N=O

Specifications

Melting Point 350°C (decomposition)
Physical Form Solid
Quantity 10 g
Assay Percent Range 96.0% min
Solubility Information Soluble in water,hot alcohol,liquid NH3Soluble in water,alcohol and ammonia.
Formula Weight 85.11
Grade ACS Reagent
Sensitivity Hygroscopic
Density 1.915 g/mL
Chemical Name or Material Potassium nitrite, For ACS analysis

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.