Learn More
Potassium nitrite, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Potassium nitrite is used as a food preservative, paint and coating additives and analytical reagent. It is used as a strong oxidizing agent as well as involved in the preparation of heat transfer salts. In addition, it is employed as corrosion inhibitors and anti-scaling agents.
Solubility
Soluble in water, alcohol and ammonia.
Notes
Hygroscopic. Incompatible with strong reducing agents, powdered metals, combustible materials, cyanides, ammonium salts and strong acids.
Specifications
Specifications
| Melting Point | 350°C (decomposition) |
| pH | 7.0 to 10.0 |
| Quantity | 100 g |
| UN Number | UN1488 |
| Merck Index | 14,7649 |
| Solubility Information | Soluble in water,alcohol and ammonia. |
| IUPAC Name | potassium nitrite |
| Formula Weight | 85.11 |
| Percent Purity | 97% |
| Odor | Odorless |
| Show More |
Safety and Handling
- Potassium nitrite
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- MIXTURE LIST-Contain: Potassium nitrite
RUO – Research Use Only
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.