missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrite, 97%, extra pure

Product Code. 15113908 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15113908 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15113908 Supplier Thermo Scientific Acros Supplier No. 453190010
  • ONLINE EXCLUSIVE PRICE
    £66.85
    £62.70 / 1kg
    Save £4.15
    6% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7758-09-0 | KNO2 | 85.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
SMILES [K+].[O-]N=O

Specifications

Melting Point 387.0°C
Quantity 1 kg
Assay Percent Range 97%
Linear Formula KNO2
Merck Index 15,777
Solubility Information Solubility in water: 3000g/L water (20°C). Other solubilities: 413 g/100mL water (100°C), slightly soluble in alcohol, soluble in hot alcohol, very soluble in liquid nh3
IUPAC Name potassium nitrite
Formula Weight 85.1
Percent Purity 97%
Grade Extra Pure
Chemical Name or Material Potassium nitrite, Extra Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium nitrite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Potassium nitrite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.