missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, ≥96.0%, ACS Reagent, Solstice

Product Code. 15663190 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15663190 500 g Poly Bottle 500g
15653190 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15663190 Supplier Honeywell-Fluka Supplier No. 310484500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥96.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
IUPAC Name potassium;nitrite
SMILES [K+].[O-]N=O

Specifications

Melting Point 350°C
Quantity 500 g
UN Number UN1488
Formula Weight 85.1g/mol
Percent Purity ≥96.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.920g/cm3
Chemical Name or Material Potassium Nitrite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.