missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate, TraceSELECT™, ≥99.995% (metals basis), Honeywell™ Fluka™

Product Code. 15675970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15675970 25 g Poly bottle 25g
15665970 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15675970 Supplier Honeywell-Fluka Supplier No. 6042925G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Melting Point 335°C (1013hPa)
Color Colorless
pH 5.5 to 7.5
Physical Form Crystalline
Quantity 25 g
UN Number UN1486
Formula Weight 101.1g/mol
Percent Purity ≥99.995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly bottle
Density 2.11g/cm3
Chemical Name or Material Potassium nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.