missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate, Technical, Fisher Chemical™

Product Code. 10598440 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
3 kg
Unit Size:
1kg
3kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10598440 1 kg 1kg
10578630 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10598440 Supplier Fisher Chemical Supplier No. P/6040/60
  • ONLINE EXCLUSIVE PRICE
    £54.93
    £39.90 / 1kg
    Save £15.03
    27% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400°C
Melting Point 334°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 1 kg
IUPAC Name potassium nitrate
Formula Weight 101.1g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Potassium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.