missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Solstice
Shop All Honeywell Fluka ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 100 g |
| UN Number | 1486 |
| IUPAC Name | potassium;nitrate |
| Formula Weight | 101.1g/mol |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. |
| Chemical Name or Material | Potassium Nitrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
Product Identifier
- Potassium nitrate
- Warning
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?