missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrate, Puriss., meets analytical spec. of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%, Honeywell Fluka™

Puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Brand: Honeywell-Fluka 12648-25KG
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| 7757-79-1 | |
| 25kg | |
| KNO3 | |
| MFCD00011409 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| [N+](=O)([O-])[O-].[K+] | |
| 101.102 | |
| CHEBI:63043 | |
| 99.0 to 100.5% | |
| Paper / Poly Bag | |
| Potassium nitrate, Puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5% |
| 100.0000% | |
| 99 to 100.5% | |
| KNO3 | |
| UN1486 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium;nitrate | |
| 24434 | |
| 101 | |
| puriss. | |
| 2.11 g/cm3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?