missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, Puratronic™, 99.995% (metals basis)

Product Code. 11386987
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386987 50 g 50g
11366987 5 g 5g
11376987 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386987 Supplier Thermo Scientific Alfa Aesar Supplier No. 010844.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7757-79-1 | IK | 166.003 g/mol

Potassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
MDL Number MFCD00011409
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346

Specifications

Boiling Point 400°C (decomposition)
Melting Point 334°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.998% (metals basis)
Molecular Formula IK
Solubility Information Moderately soluble in water,and insoluble in alcohol. Soluble in glycerol and ammonia.
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
Formula Weight 101.11
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline powder
Vapor Pressure 1.33 hpa (1mm Hg) at 745°C
Density 2.109
Chemical Name or Material Potassium nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.