missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99+%, ACS reagent, Thermo Scientific™

£18.50 - £39.60

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.1
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium;nitrate
SMILES [N+](=O)([O-])[O-].[K+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10023353
SDS View Documents Promotion Details
Thermo Scientific Acros
424151000
100g Glass bottle
ONLINE EXCLUSIVE PRICE
£19.10
£18.50 / 100g
Save £0.60
3% Off
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
 
 Restricted Item
10557372
SDS View Documents Promotion Details
Thermo Scientific Acros
424155000
500g Glass bottle
£39.60
500g
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7757-79-1
101.1
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[N+](=O)([O-])[O-].[K+]

Specifications

7757-79-1
KNO3
MFCD00011409
05,561
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.1
ACS Reagent
Potassium nitrate, crystals
99+%
KNO3
1486
15, 7769
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol
[N+](=O)([O-])[O-].[K+]
101.1
CHEBI:63043
99+%
2.1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 231-818-8

missing translation for 'rtecsNumber' : TT3700000

missing translation for 'tsca' : TSCA

Product Identifier
  • Potassium nitrate

Signal Word
  • Warning

Hazard Category
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.