missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate, ≥99.0%, ACS Reagent, Honeywell Fluka™

Product Code. p-7120716 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15612110

Brand: Honeywell-Fluka 221295100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Quantity 100 g
UN Number UN1486
IUPAC Name potassium nitrate
Formula Weight 101.1g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Nitrate
compliance-icons
Product Identifier
  • Potassium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.