missing translation for 'onlineSavingsMsg'
Learn More

Potassium molybdenum oxide, 95%

Product Code. 11478630
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478630 100 g 100g
11488630 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478630 Supplier Thermo Scientific Alfa Aesar Supplier No. 048112.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-49-6 | K2MoO4 | 238.14 g/mol

Used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-49-6
Molecular Formula K2MoO4
Molecular Weight (g/mol) 238.14
MDL Number MFCD00011370
InChI Key NUYGZETUIUXJCO-UHFFFAOYSA-N
Synonym potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
PubChem CID 6093816
IUPAC Name dipotassium;dioxido(dioxo)molybdenum
SMILES [K+].[K+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 919°C
Physical Form Powder
Quantity 100 g
Merck Index 14,7647
Solubility Information Soluble in water; Insoluble in alcohol
Formula Weight 238.14
Percent Purity 95%
Odor Odorless
Sensitivity Hygroscopic
Density 2.3 g/mL
Chemical Name or Material Potassium molybdenum oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.