missing translation for 'onlineSavingsMsg'
Learn More

Potassium molybdate, 95%

Product Code. 12694987 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12694987 500 g 500g
12684987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12694987 Supplier Thermo Scientific Acros Supplier No. 393235000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-49-6 | K2MoO4 | 238.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-49-6
Molecular Formula K2MoO4
Molecular Weight (g/mol) 238.14
MDL Number MFCD00011370
InChI Key NUYGZETUIUXJCO-UHFFFAOYSA-N
Synonym potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
PubChem CID 6093816
IUPAC Name dipotassium;dioxido(dioxo)molybdenum
SMILES [K+].[K+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 919.0°C
Quantity 500 g
Assay Percent Range 94% min
Linear Formula K2MoO4
Merck Index 15,7768
Formula Weight 238.14
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Potassium molybdate
compliance-icons
Product Identifier
  • Potassium molybdate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.