missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals

Product Code. 10733431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10733431 500 mL Glass bottle 500mL
11954611 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10733431 Supplier Thermo Scientific Acros Supplier No. 240605000
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 865-33-8 | Molecular Formula: CH3KO | Molecular Weight: 70.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-33-8 , 67-56-1 , 108-88-3
Molecular Formula CH3KO
Molecular Weight (g/mol) 70.132
InChI Key BDAWXSQJJCIFIK-UHFFFAOYSA-N
Synonym potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
PubChem CID 23664618
IUPAC Name potassium;methanolate
SMILES C[O-].[K+]

Specifications

Color Colorless
Linear Formula CH3OK
Concentration 0.1N, exact strength on the certificate of analysis
Density 0.8500g/mL
Solubility Information Solubility in water: may decompose.
Formula Weight 70.13
Grade Pure
Packaging Glass bottle
Specific Gravity 0.85
Quantity 500 mL
Flash Point 7°C
Physical Form Liquid
Chemical Name or Material Potassium methoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium methoxide, 0.1N solution in toluene / methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.