missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, 90+%

Product Code. 11337847
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337847 250 g 250g
11347847 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337847 Supplier Thermo Scientific Alfa Aesar Supplier No. 014261.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 865-33-8 | CH3KO | 70.132 g/mol

Potassium methoxide is used as a chemical intermediate. It is also used in biodiesel production and in fuel additives. It acts as a catalyst for the preparation of methyl formate and an alkaline catalyst for dimethyl formamide. Further, it is used as versatile reagents in chemical reactions such as condensation, esterification, alkoxylation and etherification, Claisen condensation, Wolf-Kishner reduction and Stobbe reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-33-8
Molecular Formula CH3KO
Molecular Weight (g/mol) 70.132
MDL Number MFCD00012416
InChI Key BDAWXSQJJCIFIK-UHFFFAOYSA-N
Synonym potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
PubChem CID 23664618
IUPAC Name potassium;methanolate
SMILES C[O-].[K+]

Specifications

Physical Form Powder
Quantity 250 g
Assay Percent Range >90%
Solubility Information Soluble in methanol and ethanol.
Formula Weight 70.14
Sensitivity Air and moisture sensitive
Chemical Name or Material Potassium methoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.