missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, 5% w/v in methanol, Thermo Scientific Chemicals

Product Code. 11399148
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399148 100 mL 100mL
11389148 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399148 Supplier Thermo Scientific Alfa Aesar Supplier No. 042551.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 865-33-8 | Molecular Formula: CH3KO | Molecular Weight: 70.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-33-8
Molecular Formula CH3KO
Molecular Weight (g/mol) 70.132
MDL Number MFCD00012416
InChI Key BDAWXSQJJCIFIK-UHFFFAOYSA-N
Synonym potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
PubChem CID 23664618
IUPAC Name potassium;methanolate
SMILES C[O-].[K+]

Specifications

Odor Alcohol-like
Linear Formula KOCH3
Quantity 100 mL
UN Number UN3274
Sensitivity Air & Moisture Sensitive
Formula Weight 70.14
Concentration or Composition (by Analyte or Components) 5% w/v in methanol
Physical Form Liquid
Chemical Name or Material Potassium methoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.