missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, 5% w/v in methanol, Thermo Scientific Chemicals

Catalog Number p-7052098
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
865-33-8
CH3KO
70.132
MFCD00012416
BDAWXSQJJCIFIK-UHFFFAOYSA-N
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
23664618
potassium;methanolate
C[O-].[K+]
This item is not returnable. View return policy
865-33-8
CH3KO
70.132
MFCD00012416
BDAWXSQJJCIFIK-UHFFFAOYSA-N
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
23664618
potassium;methanolate
C[O-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Alcohol-like
Linear Formula KOCH3
Quantity 25 mL
UN Number UN3274
Sensitivity Air and moisture sensitive
Formula Weight 70.14
Concentration or Composition (by Analyte or Components) 5% w/v in methanol
Physical Form Liquid
Chemical Name or Material Potassium methoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'