missing translation for 'onlineSavingsMsg'
Learn More

Potassium metabisulfite, 97%, extra pure

Product Code. 10320354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10320354 2.5 kg Plastic bottle 2.5kg
10722792 25 g Glass bottle 25g
10229560 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10320354 Supplier Thermo Scientific Acros Supplier No. 418270025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16731-55-8 | K2O5S2 | 222.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
PubChem CID 28019
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

Melting Point 150.0°C
Color White to Yellow
Physical Form Fine Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 96% min. (Iodimetry)
Linear Formula K2S2O5
Merck Index 15, 7766
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: insoluble in ethanol
Formula Weight 222.33
Percent Purity 97%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium metabisulfite, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Potassium metabisulfite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.